missing translation for 'onlineSavingsMsg'
Learn More

TrkB Antibody (8D2E8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-52523-0.025mg

 View more versions of this product

Product Code. 18669697

  • £173.00 / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TrkB Monoclonal specifically detects TrkB in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TrkB
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready
BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B
Mouse
92 kDa
0.025 mg
GPCR, Neuroendocrinology, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction
4915
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG1
Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry
8D2E8
Unconjugated
PBS with 0.05% Sodium Azide
NTRK2
Purified recombinant fragment of human NTRK2 (AA: 207-339) expressed in E. Coli.
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.