Learn More
Tris[N,N-bis(trimethylsilyl)amide]neodymium(III), 98%, Thermo Scientific Chemicals
£469.00
Chemical Identifiers
CAS | 41836-23-1 |
---|---|
Molecular Formula | C18H54N3NdSi6 |
Molecular Weight (g/mol) | 625.403 |
MDL Number | MFCD03427089 |
InChI Key | LWKIKGQVWYVALF-UHFFFAOYSA-N |
Synonym | tris n,n-bis trimethylsilyl amide neodymium iii, 1,1,1-trimethyl-n-trimethylsilyl silanamine neodymium iii salt, 2,2,6,6-tetramethyl-n,n,3,5-tetrakis trimethylsilyl-3,5-diaza-2,6-disila-4-neodymaheptan-4-amine |
PubChem CID | 4397443 |
IUPAC Name | bis(trimethylsilyl)azanide;neodymium(3+) |
SMILES | C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Nd+3] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
41836-23-1 | |
625.403 | |
LWKIKGQVWYVALF-UHFFFAOYSA-N | |
4397443 | |
C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Nd+3] |
C18H54N3NdSi6 | |
MFCD03427089 | |
tris n,n-bis trimethylsilyl amide neodymium iii, 1,1,1-trimethyl-n-trimethylsilyl silanamine neodymium iii salt, 2,2,6,6-tetramethyl-n,n,3,5-tetrakis trimethylsilyl-3,5-diaza-2,6-disila-4-neodymaheptan-4-amine | |
bis(trimethylsilyl)azanide;neodymium(3+) |
Specifications
41836-23-1 | |
Purple | |
[[(CH3)3Si]2N]3Nd | |
UN3396 | |
tris n,n-bis trimethylsilyl amide neodymium iii, 1,1,1-trimethyl-n-trimethylsilyl silanamine neodymium iii salt, 2,2,6,6-tetramethyl-n,n,3,5-tetrakis trimethylsilyl-3,5-diaza-2,6-disila-4-neodymaheptan-4-amine | |
C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Nd+3] | |
625.403 | |
625.39 | |
Powder |
156°C to 159°C | |
C18H54N3NdSi6 | |
MFCD03427089 | |
Moisture sensitive | |
LWKIKGQVWYVALF-UHFFFAOYSA-N | |
bis(trimethylsilyl)azanide;neodymium(3+) | |
4397443 | |
98% | |
Tris[N,N-bis (trimethylsilyl)amide]neodymium (III) |
Safety and Handling
GHS H Statement
H261-H228-H314-H318
In contact with water releases flammable gas.
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P223-P231+P232-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
H228-H261-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, WATER-REACTIVE, FLAMMABLE
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only