missing translation for 'onlineSavingsMsg'
Learn More

Tris[N,N-bis(trimethylsilyl)amide]neodymium(III), 98%, Thermo Scientific Chemicals

Product Code. 11438590
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438590 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11438590 Supplier Thermo Scientific Alfa Aesar Supplier No. 044395.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41836-23-1
Molecular Formula C18H54N3NdSi6
Molecular Weight (g/mol) 625.403
MDL Number MFCD03427089
InChI Key LWKIKGQVWYVALF-UHFFFAOYSA-N
Synonym tris n,n-bis trimethylsilyl amide neodymium iii, 1,1,1-trimethyl-n-trimethylsilyl silanamine neodymium iii salt, 2,2,6,6-tetramethyl-n,n,3,5-tetrakis trimethylsilyl-3,5-diaza-2,6-disila-4-neodymaheptan-4-amine
PubChem CID 4397443
IUPAC Name bis(trimethylsilyl)azanide;neodymium(3+)
SMILES C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Nd+3]

Specifications

Melting Point 156°C to 159°C
Color Purple
Linear Formula [[(CH3)3Si]2N]3Nd
Quantity 5 g
UN Number UN3396
Sensitivity Moisture sensitive
Formula Weight 625.39
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Tris[N,N-bis (trimethylsilyl)amide]neodymium (III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.