missing translation for 'onlineSavingsMsg'
Learn More

Tris[N,N-bis(trimethylsilyl)amide]erbium(III), 98%

Product Code. 11448620
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448620 1 g 1g
11438620 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448620 Supplier Thermo Scientific Alfa Aesar Supplier No. 044421.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103457-72-3 | C18H54ErN3Si6 | 648.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103457-72-3
Molecular Formula C18H54ErN3Si6
Molecular Weight (g/mol) 648.42
MDL Number MFCD00271002
InChI Key ZVNGWTZZRANJAO-UHFFFAOYSA-N
Synonym bis trimethylsilyl azanide; erbium 3+, tris n,n-bis trimethylsilyl amide erbium iii, acmc-20al2x, erbium iii bis trimethylsilyl amide, tris bis trimethylsilyl amino erbium, erbium tris bis trimethylsilyl amide, erbium 3+ tris bis trimethylsilyl azanide, erbium tris trimethyl-n-trimethylsilyl silanaminide, silanamine,1,1,1-trimethyl-n-trimethylsilyl-, erbium 3+ salt 3:1
PubChem CID 6098739
IUPAC Name bis(trimethylsilyl)azanide;erbium(3+)
SMILES C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Er+3]

Specifications

Melting Point 166°C
Color Pink
Linear Formula [[(CH3)3Si]2N]3Er
Quantity 1 g
UN Number UN3396
Sensitivity Moisture sensitive
Formula Weight 648.41
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Tris[N,N-bis (trimethylsilyl)amide]erbium (III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.