missing translation for 'onlineSavingsMsg'
Learn More

Tris[N,N-bis(trimethylsilyl)amide]erbium(III), 98%

Product Code. 11438620
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11438620

Brand: Thermo Scientific Alfa Aesar 044421.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103457-72-3 | C18H54ErN3Si6 | 648.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103457-72-3
Molecular Formula C18H54ErN3Si6
Molecular Weight (g/mol) 648.42
MDL Number MFCD00271002
InChI Key ZVNGWTZZRANJAO-UHFFFAOYSA-N
Synonym bis trimethylsilyl azanide; erbium 3+, tris n,n-bis trimethylsilyl amide erbium iii, acmc-20al2x, erbium iii bis trimethylsilyl amide, tris bis trimethylsilyl amino erbium, erbium tris bis trimethylsilyl amide, erbium 3+ tris bis trimethylsilyl azanide, erbium tris trimethyl-n-trimethylsilyl silanaminide, silanamine,1,1,1-trimethyl-n-trimethylsilyl-, erbium 3+ salt 3:1
PubChem CID 6098739
IUPAC Name bis(trimethylsilyl)azanide;erbium(3+)
SMILES C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Er+3]

Specifications

Melting Point 166°C
Color Pink
Linear Formula [[(CH3)3Si]2N]3Er
Quantity 5 g
UN Number UN3396
Sensitivity Moisture sensitive
Formula Weight 648.41
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Tris[N,N-bis (trimethylsilyl)amide]erbium (III)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.