missing translation for 'onlineSavingsMsg'
Learn More

Tris(ethylenediamine)iron(III) sulfate, Thermo Scientific™

Product Code. 11347408
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347408 50 g 50g
11357408 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347408 Supplier Thermo Scientific Alfa Aesar Supplier No. 031123.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54398-24-2
Molecular Formula C12H48Fe2N12O12S3
Molecular Weight (g/mol) 760.46
MDL Number MFCD00035682
InChI Key VXCQKPYJBNCKLI-UHFFFAOYSA-H
Synonym ferric ethylene diamine sulfate, tris ethylenediamine iron iii sulfate
PubChem CID 19601230

Specifications

Quantity 50 g
Formula Weight 760.38
Physical Form Powder
Chemical Name or Material Tris(ethylenediamine)iron(III) sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.