CAS RN 54398-24-2
IUPAC Name:
diiron(3+) hexakis(ethane-1,2-diamine) trisulfate
Synonyms:
diiron(3+) hexakis(ethylenediamine) trisulfate
Molecular Weight (g/mol):
760.46
Molecular Formula:
C12H48Fe2N12O12S3
InChi Key:
VXCQKPYJBNCKLI-UHFFFAOYSA-H
SMILES:
[Fe+3].[Fe+3].NCCN.NCCN.NCCN.NCCN.NCCN.NCCN.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
1
–
1
of
1
results
1
Tris(ethylenediamine)iron(III) sulfate, Thermo Scientific™
CAS: 54398-24-2 Molecular Formula: C12H48Fe2N12O12S3 Molecular Weight (g/mol): 760.46 MDL Number: MFCD00035682 InChI Key: VXCQKPYJBNCKLI-UHFFFAOYSA-H Synonym: ferric ethylene diamine sulfate,tris ethylenediamine iron iii sulfate PubChem CID: 19601230