missing translation for 'onlineSavingsMsg'
Learn More

Tris(dimethylamino)gallium(III) dimer, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11358398
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358398 2.5 g 2.5g
11368398 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358398 Supplier Thermo Scientific Alfa Aesar Supplier No. 040443.27
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Gallium nitride film precursor

Gallium nitride film precursor. Used as Reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57731-40-5
Molecular Formula C12H36Ga2N6
Molecular Weight (g/mol) 403.908
MDL Number MFCD00798535
InChI Key OHLCFMPFTXQSJZ-UHFFFAOYSA-N
Synonym gallium dimethylamide, tris dimethylamido gallium iii 98, bis tris dimethylamino gallium dimer
PubChem CID 16717646
IUPAC Name N-[bis(dimethylamino)gallanyl]-N-methylmethanamine
SMILES CN(C)[Ga](N(C)C)N(C)C.CN(C)[Ga](N(C)C)N(C)C

Specifications

Melting Point 91°C
Color White to Yellow
Boiling Point 125°C (0.01 mmHg)(sublimation)
Packaging Ampule
Linear Formula Ga2[N(CH3)2]6
Quantity 2.5 g
UN Number UN3131
Sensitivity Moisture sensitive
Solubility Information Miscible with Water.
Formula Weight 403.9
Percent Purity 99.9%
Physical Form Chunks and/or Powder
Assay (metals basis)
Chemical Name or Material Tris(dimethylamino)gallium(III) dimer
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.