accessibility menu, dialog, popup

UserName

H3CNCH3GaNCH3CH3NCH3H3CH3CNCH3GaNCH3CH3NCH3H3C

CAS RN 57731-40-5

IUPAC Name: bis([bis(dimethylamino)gallanyl]dimethylamine)
Synonyms: bis(tris(dimethylamino)gallium dimer)
Molecular Weight (g/mol): 403.91
Molecular Formula: C12H36Ga2N6
InChi Key: OHLCFMPFTXQSJZ-UHFFFAOYSA-N
SMILES: CN(C)[Ga](N(C)C)N(C)C.CN(C)[Ga](N(C)C)N(C)C
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)