missing translation for 'onlineSavingsMsg'
Learn More

Tris(dimethylamino)gallium(III) dimer, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11368398
Click to view available options
Quantity:
500 mg
2.5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Gallium nitride film precursor

Gallium nitride film precursor. Used as Reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57731-40-5
Molecular Formula C12H36Ga2N6
Molecular Weight (g/mol) 403.908
MDL Number MFCD00798535
InChI Key OHLCFMPFTXQSJZ-UHFFFAOYSA-N
Synonym gallium dimethylamide, tris dimethylamido gallium iii 98, bis tris dimethylamino gallium dimer
PubChem CID 16717646
IUPAC Name N-[bis(dimethylamino)gallanyl]-N-methylmethanamine
SMILES CN(C)[Ga](N(C)C)N(C)C.CN(C)[Ga](N(C)C)N(C)C

Specifications

Melting Point 91°C
Color White to Yellow
Boiling Point 125°C (0.01 mmHg)(sublimation)
Packaging Ampule
Linear Formula Ga2[N(CH3)2]6
Quantity 500 mg
UN Number UN3131
Sensitivity Moisture Sensitive
Formula Weight 403.9
Percent Purity 99.9%
Physical Form Chunks and/or Powder
Assay (metals basis)
Chemical Name or Material Tris(dimethylamino)gallium(III) dimer
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'