missing translation for 'onlineSavingsMsg'
Learn More

Tris(cyclopentadienyl)praseodymium(III), 99.9% (REO), Thermo Scientific™

Product Code. 11308218
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
This item is not returnable. View return policy

Product Code. 11308218

Brand: Thermo Scientific Alfa Aesar 039428.02

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11077-59-1
Molecular Formula C15H15Pr
Molecular Weight (g/mol) 336.193
MDL Number MFCD00145478
InChI Key YFBJZFPKCQMKSJ-UHFFFAOYSA-N
Synonym tris cyclopentadienyl praseodymium, acmc-1byo4
PubChem CID 18417973
IUPAC Name cyclopenta-1,3-diene;praseodymium(3+)
SMILES C1C=CC=[C-]1.C1C=CC=[C-]1.C1C=CC=[C-]1.[Pr+3]

Specifications

Melting Point 427°C (decomposition)
Boiling Point 220°C (0.01 mmHg)(sublimation)
Packaging Ampule
Quantity 0.5 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Formula Weight 336.2
Percent Purity 99.9%
Physical Form Powder
Assay (REO)
Chemical Name or Material Tris(cyclopentadienyl)praseodymium(III)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.