accessibility menu, dialog, popup

UserName

CAS RN 11077-59-1

Pr

CAS RN 11077-59-1

IUPAC Name: tris(cyclopenta-2,4-dien-1-yl)praseodymium
Synonyms: tris(cyclopenta-2,4-dien-1-yl)praseodymium
Molecular Weight (g/mol): 336.19
Molecular Formula: C15H15Pr
InChi Key: GXWCTXIZAHJGOU-UHFFFAOYSA-N
SMILES: C1=CC(C=C1)[Pr](C1C=CC=C1)C1C=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)