missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)holmium(III), Thermo Scientific™

Product Code. 11357748
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357748 1 g 1g
11367748 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357748 Supplier Thermo Scientific Alfa Aesar Supplier No. 033531.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15522-73-3
Molecular Formula C33H57HoO6
Molecular Weight (g/mol) 714.743
MDL Number MFCD00070457
InChI Key BWNBYURALNPFHU-LWTKGLMZSA-K
Synonym holmium iii-dpm, holmium 2,2,6,6-tetramethyl-3,5-heptanedionate, ho tmhd 3, tris 2,2,6,6-tetramethyl-3,5-heptanedionato holmium iii, tris 2,2,6,6-tetramethyl-3,5-heptanedionato holmium, 4z-5-bis 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy holmio oxy-2,2,6,6-tetramethylhept-4-en-3-one
PubChem CID 134067810
IUPAC Name holmium(3+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
SMILES CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Ho+3]

Specifications

Melting Point 179°C to 183°C
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 714.73
Physical Form Crystalline
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)holmium(III)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.