CAS RN 15522-73-3
IUPAC Name:
holmium(3+) tris(2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate)
Synonyms:
holmium(3+) tris(2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate)
Molecular Weight (g/mol):
714.74
Molecular Formula:
C33H57HoO6
InChi Key:
BWNBYURALNPFHU-UHFFFAOYSA-K
SMILES:
[Ho+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C
1
–
1
of
1
results
1
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)holmium(III), Thermo Scientific™
CAS: 15522-73-3 Molecular Formula: C33H57HoO6 Molecular Weight (g/mol): 714.743 MDL Number: MFCD00070457 InChI Key: BWNBYURALNPFHU-LWTKGLMZSA-K Synonym: holmium iii-dpm,holmium 2,2,6,6-tetramethyl-3,5-heptanedionate,ho tmhd 3,tris 2,2,6,6-tetramethyl-3,5-heptanedionato holmium iii,tris 2,2,6,6-tetramethyl-3,5-heptanedionato holmium,4z-5-bis 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy holmio oxy-2,2,6,6-tetramethylhept-4-en-3-one PubChem CID: 134067810 IUPAC Name: holmium(3+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate SMILES: CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].[Ho+3]