missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99%

Product Code. 11375048
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11375048 10 g 10g
11365048 0.5 g 0.5g
11355048 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11375048 Supplier Thermo Scientific Alfa Aesar Supplier No. 043993.09
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Vapor deposition | CAS: 14877-41-9 | C33H57CoO6 | 608.75 g/mol

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III) is used precursor for aerosol-assisted CVD of mixed-conducting ceramic films of Sr-Co-Fe perovskite phases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14877-41-9
Molecular Formula C33H57CoO6
Molecular Weight (g/mol) 608.75
MDL Number MFCD00233617,MFCD00233617
InChI Key UJRJNARBHKKZST-UHFFFAOYSA-K
Synonym cobalt tris 2,2,6,6-tetramethyl-3,5-heptanedionate, cobalt 3+ tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
PubChem CID 131675252
SMILES [Co+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C

Specifications

Melting Point 254°C to 256°C
Color Green
Boiling Point 120°C (0.5 mmHg)(sublimation)
Quantity 10 g
Solubility Information Insoluble in water.
IUPAC Name cobalt(3+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
Formula Weight 608.72
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt (III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.