accessibility menu, dialog, popup

UserName

CH3H3CCH3OCH3H3CCH3OCH3H3CCH3OCH3H3CCH3OCH3H3CCH3OCH3H3CCH3OCo(E/Z)(E/Z)(E/Z)

CAS RN 14877-41-9

IUPAC Name: cobalt(3+) tris(2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate)
Synonyms: cobalt(3+) tris(2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate)
Molecular Weight (g/mol): 608.75
Molecular Formula: C33H57CoO6
InChi Key: UJRJNARBHKKZST-UHFFFAOYSA-K
SMILES: [Co+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C
Boiling Point 
  • (5)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)