missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99.9% (metals basis), Thermo Scientific™

Product Code. 11339667
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339667 1 g 1g
11329667 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339667 Supplier Thermo Scientific Alfa Aesar Supplier No. 041280.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14877-41-9
Molecular Formula C33H57CoO6
Molecular Weight (g/mol) 608.75
MDL Number MFCD00233617,MFCD00233617
InChI Key UJRJNARBHKKZST-UHFFFAOYSA-K
Synonym cobalt tris 2,2,6,6-tetramethyl-3,5-heptanedionate, cobalt 3+ tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
PubChem CID 131675252
IUPAC Name cobalt(3+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
SMILES [Co+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C

Specifications

Melting Point 254°C to 256°C
Color Green
Boiling Point 120°C (0.5 mmHg)(sublimation)
Odor Odorless
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 608.72
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.