missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99.9% (metals basis), Thermo Scientific™

Product Code. 11329667
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329667 5 g 5g
11339667 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329667 Supplier Thermo Scientific Alfa Aesar Supplier No. 041280.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14877-41-9
Molecular Formula C33H57CoO6
Molecular Weight (g/mol) 608.75
MDL Number MFCD00233617,MFCD00233617
InChI Key UJRJNARBHKKZST-UHFFFAOYSA-K
Synonym cobalt tris 2,2,6,6-tetramethyl-3,5-heptanedionate, cobalt 3+ tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
PubChem CID 131675252
IUPAC Name cobalt(3+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
SMILES [Co+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C

Specifications

Melting Point 254°C to 256°C
Color Green
Boiling Point 120°C (0.5 mmHg)(sublimation)
Odor Odorless
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 608.72
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III)
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.