missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine

Product Code. 15436030
Change view
Click to view available options
Quantity:
100/Pk.
Format:
55mm Contact Plate
90mm Monoplate
Media Type:
Tryptone Soya Agar
Manufacturing Origin:
Germany
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format Media Type Manufacturing Origin Unit Size
15436030 100/Pk. 90mm Monoplate Tryptone Soya Agar Germany Pack of 100
15476030 100/Pk. 55mm Contact Plate Tryptone Soya Agar Germany Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436030 Supplier Thermo Scientific™ Supplier No. PO5501B
 Restricted Item
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Carry out environmental monitoring in your most regulated environments using Thermo Scientific™ Triple Wrapped Plates for passive and active air sampling applications.

Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Tryptone Soya Agar plates, complete with VHP indicator, Lecithin, Polysorbate 80, Sodium Thiosulphate and L-Histidine.

Achieve confidence in your results and ensure optimum media fertility using plates with a Vapour-phase Hydrogen Peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.

Three protective layers reduce risk of contamination:
  • Primary film with moisture control patch
  • Secondary film VHP barrier with immobilized desiccant sachet
  • Tertiary film dust cover

  • The incorporation of novel elements enhance moisture control by permitting the passage and absorption of excess moisture
     
  • Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs

Not all products are available for sale in all territories. Please inquire.
TRUSTED_SUSTAINABILITY

Specifications

Color Ivory, transparent
Content And Storage 2 to 25°C
Format 90mm Monoplate
Manufacturing Origin Germany
Media Type Tryptone Soya Agar
Product Type Agar Plate
Fill Weight 25.0g ± 5%
Packaging Type Triple Wrapped - Irradiated
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.