missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Triple Wrap Sterile Pack w/VHP Indicator Sabouraud Dextrose Agar
Description
Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Sabouraud Dextrose Agar settling plates, complete with VHP Indicator.
Achieve confidence in your results and ensure optimum media fertility using plates with a Vapour-phase Hydrogen Peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.
Three protective layers reduce risk of contamination:
Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs
Not all products are available for sale in all territories. Please inquire.
Three protective layers reduce risk of contamination:
- Primary film with moisture control patch
- Secondary film VHP barrier with immobilized desiccant sachet
- Tertiary film dust cover
Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs
Not all products are available for sale in all territories. Please inquire.
Specifications
Specifications
| Color | Ivory, transparent |
| Content And Storage | 2 to 25°C |
| Format | 55mm Contact Plate |
| Manufacturing Origin | Germany |
| Media Type | Sabouraud Dextrose Agar |
| Product Type | Agar |
| Fill Weight | 13.0g ± 0.5g |
| Packaging Type | Triple Wrapped - Irradiated |
| Quantity | 100/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?