missing translation for 'onlineSavingsMsg'
Learn More

Triphenylsulfonium hexafluoroarsenate, As 14.5% min, Thermo Scientific™

Product Code. 11317977
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317977 1 g 1g
11327977 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11317977 Supplier Thermo Scientific Alfa Aesar Supplier No. 020847.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57900-42-2
Molecular Formula C18H15AsF6S
Molecular Weight (g/mol) 452.29
MDL Number MFCD00061401
InChI Key SYENPFUVTQGHIM-UHFFFAOYSA-N
Synonym triphenylsulfonium hexafluoroarsenate, triphenylsulphonium hexafluoroarsenate 1-, sulfonium, triphenyl-, hexafluoroarsenate 1-, acmc-1b107, sulfonium, triphenyl-, hexafluoroarsenate 1-1:1, hexafluoroarsenic 1-; triphenylsulfanium, hexafluoro-??-arsanuide; triphenylsulfonium
PubChem CID 93839
IUPAC Name hexafluoroarsenic(1-);triphenylsulfanium
SMILES C1=CC=C(C=C1)[S+](C2=CC=CC=C2)C3=CC=CC=C3.F[As-](F)(F)(F)(F)F

Specifications

Melting Point 195°C to 197°C
Color White
Linear Formula (C6H5)3SAsF6
Quantity 1 g
UN Number UN1557
Sensitivity Moisture sensitive
Formula Weight 452.29
Concentration or Composition (by Analyte or Components) As 14.5% min.
Physical Form Powder
Chemical Name or Material Triphenylsulfonium hexafluoroarsenate
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.