accessibility menu, dialog, popup

UserName

FAsFFFFFS

CAS RN 57900-42-2

IUPAC Name: hexafluoro-λ⁵-arsanuide; triphenylsulfanium
Synonyms: hexafluoro-λ⁵-arsanuide; triphenylsulfonium
Molecular Weight (g/mol): 452.29
Molecular Formula: C18H15AsF6S
InChi Key: SYENPFUVTQGHIM-UHFFFAOYSA-N
SMILES: F[As-](F)(F)(F)(F)F.C1=CC=C(C=C1)[S+](C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results