missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tripeptidyl Peptidase Inhibitor

Product Code. 15476569
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 15476569

Brand: Thermo Scientific Chemicals J64760.MA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tripeptidyl peptidase Inhibitor is a serine protease inhibitor which irreversibly inhibits TPPII (tripeptidyl peptidase II), a giant protease which may substitute for some proteasome functions, and reversibly inhibits Cln (TPPI). AAF-CMK shows only marginal inhibition of the chymotrypsin-like activity of the proteasome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A serine protease inhibitor that acts as a potent, selective, dose-dependent, irreversible inhibitor of tripeptidylpeptidase II
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 184901-82-4
Molecular Formula C16H22ClN3O3, C18H23ClF3N3O5
Synonym H-AAF-CMK; TPPII Inhibitor

Specifications

Quantity 10 mg
Description Off-white
Content And Storage -30°C to -10°C
For Use With (Application) A serine protease inhibitor that acts as a potent, selective, dose-dependent, irreversible inhibitor of tripeptidylpeptidase II
Formula Weight 453.90g/mol
Sensitivity Light Sensitive
Product Type Signal Transduction Reagent
Form Solid
InChI Key ZFZXFWAYCYJFIZ-UHFFFAOYNA-N
SMILES OC(=O)C(F)(F)F.CC(N)C(=O)NC(C)C(=O)NC(CC1=CC=CC=C1)C(=O)CCl
IUPAC Name 2-amino-N-{1-[(4-chloro-3-oxo-1-phenylbutan-2-yl)carbamoyl]ethyl}propanamide; trifluoroacetic acid
Molecular Weight (g/mol) 453.84
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.