missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tripeptidyl-Peptidase I/TPP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | Tripeptidyl-Peptidase I/TPP1 |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18667075
|
Novus Biologicals
NBP2-48820-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18666685
|
Novus Biologicals
NBP2-48820 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tripeptidyl-Peptidase I/TPP1 Polyclonal antibody specifically detects Tripeptidyl-Peptidase I/TPP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Tripeptidyl-Peptidase I/TPP1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS (pH 7.2), 40% Glycerol | |
| 1200 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Cell growth-inhibiting gene 1 protein, ceroid-lipofuscinosis, neuronal 2, late infantile (Jansky-Bielschowsky disease), CLN2EC 3.4.14.9, growth-inhibiting protein 1, LPIC, lysosomal pepstatin insensitive protease, Lysosomal pepstatin-insensitive protease, MGC21297, TPP-1, TPP-I, Tripeptidyl aminopeptidase, tripeptidyl peptidase I, tripeptidyl-peptidase 1, Tripeptidyl-peptidase I | |
| This antibody was developed against a recombinant protein corresponding to amino acids: SSLRQRPEPQVTGTVGLHLGVTPSVIRKRYNLTSQDVGSGTSNNSQACAQFLEQYFHDSDLAQFMRLFGGNFAHQASVARVV | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title