missing translation for 'onlineSavingsMsg'
Learn More

(Trimethylsilylmethyl)triphenylphosphonium iodide, 98%, Thermo Scientific™

Product Code. 11380214
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11380214 1 g 1g
11390214 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11380214 Supplier Thermo Scientific Alfa Aesar Supplier No. L00788.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3739-98-8
Molecular Formula C22H26IPSi
Molecular Weight (g/mol) 476.41
MDL Number MFCD00051853
InChI Key PPORXBBMZZFXTB-UHFFFAOYSA-M
Synonym trimethylsilylmethyl triphenylphosphonium iodide, triphenyl trimethylsilyl methyl phosphonium iodide, triphenyl trimethylsilyl methyl phosphanium iodide, acmc-20aped, phosphonium,triphenyl trimethylsilyl methyl-, iodide 1:1
PubChem CID 22461619
IUPAC Name triphenyl(trimethylsilylmethyl)phosphanium;iodide
SMILES [I-].C[Si](C)(C)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∽185°C (decomposition)
Quantity 1 g
Beilstein 4070136
Sensitivity Light and moisture sensitive
Formula Weight 476.41
Percent Purity 98%
Chemical Name or Material (Trimethylsilylmethyl)triphenylphosphonium iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.