accessibility menu, dialog, popup

UserName

CAS RN 3739-98-8

IUPAC Name: triphenyl[(trimethylsilyl)methyl]phosphanium iodide
Synonyms: triphenyl[(trimethylsilyl)methyl]phosphanium iodide
Molecular Weight (g/mol): 476.41
Molecular Formula: C22H26IPSi
InChi Key: PPORXBBMZZFXTB-UHFFFAOYSA-M
SMILES: [I-].C[Si](C)(C)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results