missing translation for 'onlineSavingsMsg'
Learn More

Trimethylaluminum, 25% w/w in hexane, Thermo Scientific™

Product Code. 11430860
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 x 500 mL
Unit Size:
100mL
2 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430860 500 mL 500mL
15461837 100 mL 100mL
11440860 4 x 500 mL 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430860 Supplier Thermo Scientific Alfa Aesar Supplier No. 042836.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-24-1
Molecular Formula C3H9Al
Molecular Weight (g/mol) 72.087
MDL Number MFCD00008252
InChI Key JLTRXTDYQLMHGR-UHFFFAOYSA-N
Synonym trimethylaluminum, trimethylaluminium, aluminum, trimethyl, trimethylalane, unii-av210lg46j, ch3 3al, aluminum trimethanide, aluminum trimethyl, trimethylaluminum solution, trimethylaluminum, elec. gr.
PubChem CID 16682925
IUPAC Name trimethylalumane
SMILES C[Al](C)C

Specifications

Flash Point −18°C (−1°F)
Quantity 500 mL
UN Number UN3399
Beilstein 3587197
Sensitivity Air and moisture sensitive
Solubility Information Soluble in aromatic,saturated aliphatic and cycloaliphatic hydrocarbons.
Formula Weight 72.09
Physical Form Liquid
Chemical Name or Material Trimethylaluminum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.