accessibility menu, dialog, popup

UserName

CH3AlCH3H3C

CAS RN 75-24-1

IUPAC Name: trimethylalumane
Synonyms: trimethylaluminium
Molecular Weight (g/mol): 72.09
Molecular Formula: C3H9Al
InChi Key: JLTRXTDYQLMHGR-UHFFFAOYSA-N
SMILES: C[Al](C)C
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)