missing translation for 'onlineSavingsMsg'
Learn More

Trimethylaluminium, 1.0M solution in heptane, AcroSeal™

Product Code. 10211373 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10211373 800 mL AcroSeal™ Glass Bottle 800mL
10153502 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10211373 Supplier Thermo Scientific Acros Supplier No. 189278000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-24-1 | C3H9Al | 72.087 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-24-1 , 142-82-5
Molecular Formula C3H9Al
Molecular Weight (g/mol) 72.087
MDL Number MFCD00008252
InChI Key JLTRXTDYQLMHGR-UHFFFAOYSA-N
Synonym trimethylaluminum, trimethylaluminium, aluminum, trimethyl, trimethylalane, unii-av210lg46j, ch3 3al, aluminum trimethanide, aluminum trimethyl, trimethylaluminum solution, trimethylaluminum, elec. gr.
PubChem CID 16682925
IUPAC Name trimethylalumane
SMILES C[Al](C)C

Specifications

Color Colorless
Boiling Point 126.0°C
Melting Point 15.0°C
Linear Formula (CH3)3Al
Fieser 08,506; 14,46; 17,372
Density 0.6880g/mL
Merck Index 15, 9879
Solubility Information Solubility in water: reacts. Other solubilities: miscible with saturated aliphatic and aromatic, hydrocarbons
Formula Weight 72.09
Percent Purity 9.5 to 10.5 wt% (on Al basis)
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.688
Viscosity 0.90 mPa.s (30°C)
Quantity 800 mL
Flash Point −4°C
Physical Form Liquid
Chemical Name or Material Trimethylaluminium, 1.0M Solution in Heptane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trimethylaluminium, 1.0M solution in heptane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.