missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Trimethoprim

Trimethoprim, CAS # 738-70-5, is a trimethoxybenzyl-pyrimidine used as an antimicrobial.

£32.40 - £312.00

Chemical Identifiers

CAS 738-70-5
Formula Weight 290.32g/mol
Synonym 2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine
Content And Storage 2°C to 8°C
Format Powder
For Use With (Application) An antibacterial and inhibitor of formylation. Dihydrofolate reductase inhibitor with selectivity for the prokaryote enzyme
Description White
Melting Point 199°C to 203°C
Solubility Trimethoprim is soluble in water at 0.4 mg/ml; It is also soluble in DMSO at 50 mg/ml and chloroform at 18.2 mg/ml
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15445319
SDS View Documents
Thermo Scientific™
J63053.03
1 g
£32.40
1g
View alternative products
Save Up to 
»
In Stock
Edge
Add to basket
 
 Restricted Item
15455319
SDS View Documents
Thermo Scientific™
J63053.06
5 g
ONLINE EXCLUSIVE PRICE
£82.40
£74.65 / 5g
Save £7.75
9% Off
View alternative products
Save Up to 
»
In Stock
Edge
Add to basket
 
 Restricted Item
15465319
SDS View Documents
Thermo Scientific™
J63053.14
25 g
£312.00
25g
View alternative products
Save Up to 
»
Estimated Shipment: 05-10-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description
• Trimethoprim is a synthetic pyrimidine analog that binds dihydrofolate-reductase and inhibits the synthesis of folic acid; when paired with sulfamethoxazole, its activity is enhanced
• Trimethoprim has antibacterial and antiprotozoal properties
Applications
• Trimethoprim inhibits the conversion of para-aminobenzoic acid (PABA) to folic acid near the end of the synthesis pathway and can be used to study related effects
• Trimethoprim has been radiolabeled and used to trace routes of infection, and it also has been used experimentally as a fluorescent labeler to quantify dihydrofolate-reductase expression
Specifications

Chemical Identifiers

738-70-5
2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine
Powder
White
Trimethoprim is soluble in water at 0.4 mg/ml; It is also soluble in DMSO at 50 mg/ml and chloroform at 18.2 mg/ml
290.32g/mol
2°C to 8°C
An antibacterial and inhibitor of formylation. Dihydrofolate reductase inhibitor with selectivity for the prokaryote enzyme
199°C to 203°C

Specifications

White
Powder
738-70-5
290.32g/mol
199°C to 203°C
625127
2°C to 8°C
An antibacterial and inhibitor of formylation. Dihydrofolate reductase inhibitor with selectivity for the prokaryote enzyme
C14H18N4O3
2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine
Trimethoprim is soluble in water at 0.4 mg/ml; It is also soluble in DMSO at 50 mg/ml and chloroform at 18.2 mg/ml
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 212-006-2

Recommended Storage : Keep cold

Product Identifier
  • Trimethoprim

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contain: Trimethoprim

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.