missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Trimethoprim

Trimethoprim, CAS # 738-70-5, is a trimethoxybenzyl-pyrimidine used as an antimicrobial.

Brand:  Thermo Scientific™ J63053.14

Quantity 25 g

 View more versions of this product

Product Code. 15465319

  • £312.00 / 25g
 Restricted Item
Estimated Shipment: 05-10-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description
• Trimethoprim is a synthetic pyrimidine analog that binds dihydrofolate-reductase and inhibits the synthesis of folic acid; when paired with sulfamethoxazole, its activity is enhanced
• Trimethoprim has antibacterial and antiprotozoal properties
Applications
• Trimethoprim inhibits the conversion of para-aminobenzoic acid (PABA) to folic acid near the end of the synthesis pathway and can be used to study related effects
• Trimethoprim has been radiolabeled and used to trace routes of infection, and it also has been used experimentally as a fluorescent labeler to quantify dihydrofolate-reductase expression
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

738-70-5
2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine
Powder
White
Trimethoprim is soluble in water at 0.4 mg/ml; It is also soluble in DMSO at 50 mg/ml and chloroform at 18.2 mg/ml
290.32g/mol
2°C to 8°C
An antibacterial and inhibitor of formylation. Dihydrofolate reductase inhibitor with selectivity for the prokaryote enzyme
199°C to 203°C

Specifications

25 g
2°C to 8°C
An antibacterial and inhibitor of formylation. Dihydrofolate reductase inhibitor with selectivity for the prokaryote enzyme
C14H18N4O3
2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine
Trimethoprim is soluble in water at 0.4 mg/ml; It is also soluble in DMSO at 50 mg/ml and chloroform at 18.2 mg/ml
White
Powder
738-70-5
290.32g/mol
199°C to 203°C
625127
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.