missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£222.00 - £443.00
Specifications
| Antigen | GERP |
|---|---|
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18475481
|
Novus Biologicals
NBP1-89776-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18727353
|
Novus Biologicals
NBP1-89776 |
0.1 mL |
£443.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRIM8 Polyclonal specifically detects TRIM8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| GERP | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q9BZR9 | |
| 81603 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QDIEDQLYKLESDKRLVEEKVNQLKEEVRLQYEKLHQLLDEDLRQTVEVLDKAQAKFCSENAAQALHLGERMQEAKKLLGS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockdown Validated | |
| Polyclonal | |
| Rabbit | |
| Zinc Finger | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EC 6.3.2.-, GERPRING finger protein 27glioblastoma expressed ring finger protein, Glioblastoma-expressed RING finger protein, RNF27probable E3 ubiquitin-protein ligase TRIM8, tripartite motif containing 8, tripartite motif protein TRIM8, tripartite motif-containing 8, Tripartite motif-containing protein 8 | |
| TRIM8 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title