missing translation for 'onlineSavingsMsg'
Learn More

TRIM59 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen TRIM59
Concentration 800 μg/mL
Applications Western Blot, Immunofluorescence, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17291406
View Documents
Proteintech
28575-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17281406
View Documents
Proteintech
28575-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TRIM59 belongs to the tripartite motif (TRIM) family. It functions as E3 ubiquitin ligases or an adaptor protein, and plays important roles in various types of human cancers.

Specifications

Specifications

TRIM59
Western Blot, Immunofluorescence, ELISA
Unconjugated
Rabbit
286827
800 μg/mL
Polyclonal
Liquid
Human, Mouse, Rat
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.