Learn More
TRIM59 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28575-1-AP-150UL
 View more versions of this product
Product Code. 17281406
  • £349.00 / 150µL
Description
TRIM59 belongs to the tripartite motif (TRIM) family. It functions as E3 ubiquitin ligases or an adaptor protein, and plays important roles in various types of human cancers.
Specifications
Specifications
TRIM59
Polyclonal
Unconjugated
150 μL
286827
Liquid
Western Blot, Immunofluorescence, ELISA
800 μg/mL
Rabbit
Primary
Human, Mouse, Rat