missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetaldehyde hydrate, tech., ca 75% in water

Product Code. p-7021056
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11420763

Brand: Thermo Scientific Alfa Aesar A13234.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 421-53-4 | C2HF3O | 98.02 g/mol

One general application of trifluoroacetaldehyde is the preparation of trifluoroethylamino derivatives via reductive amination reaction. This synthesis includes the formation of the corresponding N,O-acetal intermediates followed by their reduction using NaBH4 or 2-picoline borane complex, affording the target trifluoroethylamino compounds in moderate to good yields. Another general application of trifluoroacetaldehyde is the synthesis of chiral α-substituted trifluoroethylamino compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
One general application of trifluoroacetaldehyde is the preparation of trifluoroethylamino derivatives via reductive amination reaction. This synthesis includes the formation of the corresponding N,O-acetal intermediates followed by their reduction using NaBH4 or 2-picoline borane complex, affording the target trifluoroethylamino compounds in moderate to good yields. Another general application of trifluoroacetaldehyde is the synthesis of chiral α-substituted trifluoroethylamino compounds.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Keep in cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 421-53-4
Molecular Formula C2HF3O
Molecular Weight (g/mol) 98.02
MDL Number MFCD00042444
InChI Key JVTSHOJDBRTPHD-UHFFFAOYSA-N
Synonym tfaih, fluoral hydrate, 1,1-ethanediol, 2,2,2-trifluoro, fluoral, hydrate, 2,2,2-trifluoro-1,1-ethanediol, acetaldehyde, trifluoro-, hydrate, 2,2,2-trifluoroethanediol, fluoral hydrate;, snpljqhjptadtaeup@, acetaldehyde, hydrate
PubChem CID 67901
IUPAC Name 2,2,2-trifluoroethane-1,1-diol
SMILES FC(F)(F)C=O

Specifications

Density 1.44
Boiling Point 104°C to 106°C
Flash Point None
Linear Formula CF3CH(OH)2
Quantity 10 g
UN Number UN1989
Solubility Information Soluble in water.
Formula Weight 116.04 (Anhydrous)
Percent Purity 75%
Grade Technical
Concentration or Composition (by Analyte or Components) ≈75% in water
Chemical Name or Material Trifluoroacetaldehyde hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.