accessibility menu, dialog, popup

UserName

OFFF

CAS RN 421-53-4

IUPAC Name: 2,2,2-trifluoroethane-1,1-diol
Synonyms: trifluoroacetaldehyde hydrate
Molecular Weight (g/mol): 116.04
Molecular Formula: C2H3F3O2
InChi Key: VGJWVEYTYIBXIA-UHFFFAOYSA-N
SMILES: OC(O)C(F)(F)F
Boiling Point 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)