Learn More
Trifluoroacetaldehyde ethyl hemiacetal, tech. 80%, Thermo Scientific Chemicals
£42.30 - £146.00
Chemical Identifiers
CAS | 433-27-2 |
---|---|
Molecular Formula | C4H7F3O2 |
Molecular Weight (g/mol) | 144.093 |
MDL Number | MFCD00000441 |
InChI Key | KLXJPQNHFFMLIG-UHFFFAOYSA-N |
Synonym | trifluoroacetaldehyde ethyl hemiacetal, fluoral ethyl hemiacetal, ethanol, 1-ethoxy-2,2,2-trifluoro, trifluoroacetaldehyde hemiethylacetal, 1-ethoxy-2,2,2-trifluoro-ethanol, trifluoroacetaldehyde ethylhemiacetal, 1-ethoxy-2,2,2-trifluoroethan-1-ol, pubchem12643, acmc-1aj38, 1-ethoxy-2,2-trifluoroethanol |
PubChem CID | 9897 |
IUPAC Name | 1-ethoxy-2,2,2-trifluoroethanol |
SMILES | CCOC(C(F)(F)F)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11371317
|
Thermo Scientific Alfa Aesar
L01554.06 |
5 g |
£42.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11381317
|
Thermo Scientific Alfa Aesar
L01554.14 |
25 g |
£146.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
433-27-2 | |
144.093 | |
KLXJPQNHFFMLIG-UHFFFAOYSA-N | |
9897 | |
CCOC(C(F)(F)F)O |
C4H7F3O2 | |
MFCD00000441 | |
trifluoroacetaldehyde ethyl hemiacetal, fluoral ethyl hemiacetal, ethanol, 1-ethoxy-2,2,2-trifluoro, trifluoroacetaldehyde hemiethylacetal, 1-ethoxy-2,2,2-trifluoro-ethanol, trifluoroacetaldehyde ethylhemiacetal, 1-ethoxy-2,2,2-trifluoroethan-1-ol, pubchem12643, acmc-1aj38, 1-ethoxy-2,2-trifluoroethanol | |
1-ethoxy-2,2,2-trifluoroethanol |
Specifications
433-27-2 | |
104°C to 106°C | |
Odorless | |
1.343 | |
UN1989 | |
trifluoroacetaldehyde ethyl hemiacetal, fluoral ethyl hemiacetal, ethanol, 1-ethoxy-2,2,2-trifluoro, trifluoroacetaldehyde hemiethylacetal, 1-ethoxy-2,2,2-trifluoro-ethanol, trifluoroacetaldehyde ethylhemiacetal, 1-ethoxy-2,2,2-trifluoroethan-1-ol, pubchem12643, acmc-1aj38, 1-ethoxy-2,2-trifluoroethanol | |
CCOC(C(F)(F)F)O | |
144.093 | |
144.09 | |
Technical |
1.23 | |
39°C (102°F) | |
C4H7F3O2 | |
MFCD00000441 | |
906797 | |
KLXJPQNHFFMLIG-UHFFFAOYSA-N | |
1-ethoxy-2,2,2-trifluoroethanol | |
9897 | |
80% | |
Trifluoroacetaldehyde ethyl hemiacetal |
Safety and Handling
GHS H Statement
H226-H302-H315-H319
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P332+P313-P363-P370+P378q-P501c
H226-H302-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
missing translation for 'einecsNumber' : 207-087-6
missing translation for 'rtecsNumber' : KK9000000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only