missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetaldehyde ethyl hemiacetal, tech. 80%

Product Code. 11371317
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371317 5 g 5g
11381317 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11371317 Supplier Thermo Scientific Alfa Aesar Supplier No. L01554.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 433-27-2 | C4H7F3O2 | 144.093 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 433-27-2
Molecular Formula C4H7F3O2
Molecular Weight (g/mol) 144.093
MDL Number MFCD00000441
InChI Key KLXJPQNHFFMLIG-UHFFFAOYSA-N
Synonym trifluoroacetaldehyde ethyl hemiacetal, fluoral ethyl hemiacetal, ethanol, 1-ethoxy-2,2,2-trifluoro, trifluoroacetaldehyde hemiethylacetal, 1-ethoxy-2,2,2-trifluoro-ethanol, trifluoroacetaldehyde ethylhemiacetal, 1-ethoxy-2,2,2-trifluoroethan-1-ol, pubchem12643, acmc-1aj38, 1-ethoxy-2,2-trifluoroethanol
PubChem CID 9897
IUPAC Name 1-ethoxy-2,2,2-trifluoroethanol
SMILES CCOC(C(F)(F)F)O

Specifications

Density 1.23
Boiling Point 104°C to 106°C
Flash Point 39°C (102°F)
Odor Odorless
Refractive Index 1.343
Quantity 5 g
UN Number UN1989
Beilstein 906797
Formula Weight 144.09
Percent Purity 80%
Grade Technical
Chemical Name or Material Trifluoroacetaldehyde ethyl hemiacetal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.