missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Tricorn™ Superose™ 6 and Superose 12 Prepacked Gel Filtration Columns

Product Code. 11300212 Shop All Cytiva Products
Click to view available options
:
Superose 12, 10/300 GL; Average particle size: 11μm; Pack size: 1 piece
This item is not returnable. View return policy

Product Code. 11300212

Brand: Cytiva 17517301

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed for high performance, laboratory scale separations of proteins and other biomolecules based on size.  Cytiva Tricorn™ Superose™ 6 and Superose 12 Prepacked Gel Filtration Columns  delivers quick and reproducible separations with high resolution.

 

  • Covers wide separation range of molecular weights from 1000 up to 5million MHighly cross-linked agarose matrixSmall average bead size of 13μm (Superose 6) and 11μm (Superose 12) are designed for laboratory-scale separations with good resolving capability
  • Durable, high-performance glass columns for reliable and reproducible separations
  • Available with two different media - Superose 6 or Superose 12

     

     

  • Bed volume: 24mL
  • Bed dimension: 10 × 300mm
  • Pressure max.: 15 bar (217psi)

  • Media

  • pH stability working range: 3 to 12
  • pH stability cleaning: 1 to 14

  • Column

  • Column I.D.: 10mm
  • Bed support: filter
  • Material (bed support): polypropylene
  • Material (column hardware): borosilicate glass, PEEK, PP, EPDM and PFR
  • Material (column tube): borosilicate glass
  • Bed height with one adaptor: 279 to 309mm
  • Bed height with two adaptors: 245 to 306mm
  •  

    Superose 6 Media:

  • Average partcle size: 13μm
  • Gel filtration separations over a very broad molecular weight rang from 5000 to 5,000,000 (globular proteins)
  • Flow rate: 0.3 to 0.5mL/min.

  • Superose 12 Media:

  • Average partcle size: 11μm
  • Gel filtration separations over a very broad molecular weight rang from 1000 to 300,000 (globular proteins)
  • Flow rate of 0.5 to 1mL/min.
  •  

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M Acetic Acid, 8M Urea, 6M Guanidine Hydrochloride, 1% SDS, Organic Solvents, 0.5M NaOH (for cleaning in-place)
Product Type Superose 12 prepacked column
Bed Volume 24mL
Diameter (English) Inner 0.39 in.
Diameter (Metric) Inner 10mm
Quantity Each
Height (English) 11.81 in.
Height (Metric) 300mm
Flow Rate 0.5 to 1mL/min.
Includes Valco fittings for AKTA™ design systems and are supplied with M6 connectors for FPLC System
Material Borosilicate Glass
Max. Pressure 435psi
pH Range 3 to 12
Particle Size 11μm
Support Type Cross-linked Agarose
Content And Storage 4° to 30°C
For Use With (Application) High performance gel filtration with a broad protein fractionation range of 5000 to 5,000,000
Show More Show Less
compliance-icons
Product Identifier
  • Superose 12 10/300 GL
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.