Learn More
Cytiva Tricorn™ Superdex™ Prepacked Gel Filtration Columns
Use for laboratory-scale separations and intermediate purification based on different sizes of proteins, peptides, and other biomolecules. Cytiva Tricorn™ Superdex™ Prepacked Gel Filtration Columns are perfect for high-resolution applications.
Brand: Cytiva 28-9065-61
Code : GX
Additional Details : Weight : 1.78720kg
Description
- Fast gel filtration involving separation based on three molecular weight ranges from 100 to 600000 M
- Spherical composite of cross-linked agarose and dextran matrix
- Comes with two different bed heights of 150 and 300mm allowing greater flexibility to suit different analytical needs
- Available with three different media: Superdex Peptide, Superdex 75, and Superdex 200
Superdex Peptide:
- Bed dimension: 10 x 300mm
- Bed volume: 24mL
- Flow velocity: 0.15 to 0.6mL/min.
- Pressure max.: 18 bar (261psi)
- Separation range: 100 to 7000 (peptides)
- pH Stability working range: 1 to 14
- Column I.D.: 10mm
- Bed height with one adaptor: 279 to 309mm
- Bed height with two adaptors: 245 to 306mm
- Bed dimension: 5 x 150mm and 10 x 300mm
- Bed volume: 3 and 24mL
- Flow velocity: 0.15 to 0.6mL/min. and 0.25 to 0.75mL/min.
- Pressure max.: 15 bar (217psi)
- Separation range: 10,000 to 600,000 (globular peptides)
- pH Stability working range: 3 to 12
- Column I.D.: 5 and 10mm
- Bed height with one adaptor: 135 to 159mm (3mL column)
- Bed height with two adaptors: 108 to 156mm (3mL column)
- Bed dimension: 5 x 150mm and 10 x 300mm
- Bed volume: 3 and 24mL
- Flow velocity: 0.15 to 0.6mL/min.
- Pressure max.: 18 bar (261psi)
- Separation range: 3000 up to 70,000 (globular proteins)
- pH Stability working range: 3 to 12
- Column I.D.: 5 and 10 mm
- Bed height with one adaptor: 108 to 156mm (3mL column)
- Bed height with two adaptors: 135 to 159mm (3mL column)
Specifications
1M Acetic Acid, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol, 70% Ethanol, 1M NaOH | |
3mL | |
5mm | |
5.9 in. | |
0.15 to 0.6mL/min. | |
Borosilicate Glass | |
1 to 14 | |
Spherical composite of cross-linked Agarose and Dextran | |
Prepacked Gel Filtration Column |
Superdex™ 200 prepacked column | |
0.19 in. | |
Each | |
150mm | |
Valco fittings for AKTA™ design systems and are supplied with M6 connecto rs for FPLC System | |
15 bar | |
13μm | |
4° to 30°C | |
High-resolution, semipreparative and analytical separations of biomolecules |