missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Includes
Valco fittings for AKTAdesign systems and are supplied with M6 connectors for FPLC System.
- Fast gel filtration involving separation based on three molecular weight ranges from 100 to 600000 M
- Spherical composite of cross-linked agarose and dextran matrix
- Comes with two different bed heights of 150 and 300mm allowing greater flexibility to suit different analytical needs
- Available with three different media: Superdex Peptide, Superdex 75, and Superdex 200
Superdex Peptide:
- Bed dimension: 10 x 300mm
- Bed volume: 24mL
- Flow velocity: 0.15 to 0.6mL/min.
- Pressure max.: 18 bar (261psi)
- Separation range: 100 to 7000 (peptides)
- pH Stability working range: 1 to 14
- Column I.D.: 10mm
- Bed height with one adaptor: 279 to 309mm
- Bed height with two adaptors: 245 to 306mm
- Bed dimension: 5 x 150mm and 10 x 300mm
- Bed volume: 3 and 24mL
- Flow velocity: 0.15 to 0.6mL/min. and 0.25 to 0.75mL/min.
- Pressure max.: 15 bar (217psi)
- Separation range: 10,000 to 600,000 (globular peptides)
- pH Stability working range: 3 to 12
- Column I.D.: 5 and 10mm
- Bed height with one adaptor: 135 to 159mm (3mL column)
- Bed height with two adaptors: 108 to 156mm (3mL column)
- Bed dimension: 5 x 150mm and 10 x 300mm
- Bed volume: 3 and 24mL
- Flow velocity: 0.15 to 0.6mL/min.
- Pressure max.: 18 bar (261psi)
- Separation range: 3000 up to 70,000 (globular proteins)
- pH Stability working range: 3 to 12
- Column I.D.: 5 and 10 mm
- Bed height with one adaptor: 108 to 156mm (3mL column)
- Bed height with two adaptors: 135 to 159mm (3mL column)
Specifications
Specifications
| Buffer | 1M Acetic Acid, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol, 70% Ethanol, 1M NaOH |
| Product Type | Superdex 200 Prepacked Column |
| Bed Volume | 24 mL |
| Diameter (English) Inner | 0.39 in. |
| Diameter (Metric) Inner | 10mm |
| Quantity | Each |
| Height (English) | 11.81 in. |
| Height (Metric) | 300mm |
| Flow Rate | 0.25 to 0.75mL/min. |
| Includes | Valco fittings for AKTA™ design systems and are supplied with M6 connecto rs for FPLC System |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?