missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Tricorn Glass Tubes
Glass tubes coated with plastic film protects individuals in the laboratory from harm if the column is dropped or operated outside its specification.
Brand: Cytiva 18-1153-17
Code : GE
Additional Details : Weight : 0.22500kg
Specifications
Glass tube | |
200mm | |
Column Accesory |
10mm | |
Borosilicate glass |