missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Tributyrin Agar
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
Tributyrin Agar consists of a stable homogenate of nutrient agar and tributyrin (glyceryl tributyrate) and is used for the detection and enumeration of lipolytic (fat-splitting) bacteria and moulds.
- Tributyrin Agar consists of a stable homogenate of nutrient agar and tributyrin (glyceryl tributyrate) and is used for the detection and enumeration of lipolytic (fat-splitting) bacteria and moulds. The medium is also frequently used for the detection of the lipolytic activity of specific organisms such as staphyloccoci, clostridia, pseudomonads and flavobacteria
Specifications
Specifications
| Form | Agar |
| Manufacturing Origin | United Kingdom |
| Product Type | Broth Tube |
| Sterility | Non-sterile |
| Target Organism Class | Bacteria |
| Test Type | Microbial Detection |
| Quantity | 100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction