missing translation for 'onlineSavingsMsg'
Learn More

Tribromoacetic acid, 99%, Thermo Scientific Chemicals

Product Code. 10512731 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10512731 25 g Glass bottle 25g
10465993 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10512731 Supplier Thermo Scientific Acros Supplier No. 139300250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-96-7
Molecular Formula C2HBr3O2
Molecular Weight (g/mol) 296.73
MDL Number MFCD00004167
InChI Key QIONYIKHPASLHO-UHFFFAOYSA-N
Synonym tribromoacetic acid, acetic acid, tribromo, tribromacetic acid, unii-71ktl99qjx, ccris 7887, 71ktl99qjx, acetic acid, 2,2,2-tribromo, snpljqbkdvidtaetp@, dsstox_cid_1668, acmc-209p0y
PubChem CID 6415
IUPAC Name 2,2,2-tribromoacetic acid
SMILES C(=O)(C(Br)(Br)Br)O

Specifications

Melting Point 130°C to 133°C
Color White to Yellow
Boiling Point 245°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CBr3CO2H
Quantity 25 g
Beilstein 02, 220
Merck Index 15, 9770
Formula Weight 296.73
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material Tribromoacetic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Tribromoacetic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.