accessibility menu, dialog, popup

UserName

CAS RN 75-96-7

OOBrBrBr

CAS RN 75-96-7

IUPAC Name: tribromoacetate
Synonyms: tribromoacetate
Molecular Weight (g/mol): 295.73
Molecular Formula: C2Br3O2
InChi Key: QIONYIKHPASLHO-UHFFFAOYSA-M
SMILES: [O-]C(=O)C(Br)(Br)Br
Molecular Weight (g/mol) 
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)