missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyltin bromide, 95%, Thermo Scientific™

Product Code. 11324869
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324869 5 g 5g
11314869 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324869 Supplier Thermo Scientific Alfa Aesar Supplier No. 071185.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1461-23-0
Molecular Formula C12H27BrSn
Molecular Weight (g/mol) 369.962
MDL Number MFCD00010660
InChI Key FVRKTAOFDKFAMI-UHFFFAOYSA-M
Synonym tributyltin bromide, tri-n-butyltin bromide, bromotributyltin, stannane, tributylbromo, tri-n-butylbromotin, tributyltinbromide, bu3snbr, bromo tributyl stannane, acmc-20ajf5, tri-n-butyl tin bromide
PubChem CID 305767
IUPAC Name bromo(tributyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)Br

Specifications

Density 1.338 g/mL
Boiling Point 163°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.507
Quantity 5 g
UN Number UN2788
Sensitivity Moisture sensitive
Formula Weight 369.96
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tri-n-butyltin bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.