missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butylborane, 1M soln. in THF, Thermo Scientific Chemicals

Product Code. 15435247
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435247 500 mL 500mL
15425247 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15435247 Supplier Thermo Scientific Alfa Aesar Supplier No. H32518.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butylborane is used as a reactant for addition reactions for synthesis of phenyltrialkylborate salts, boron-based multicomponent reactions for generation of substituted aziridines, oxazolidines, and pyrrolidines, Suzuki-Miyaura cross coupling of halopurines with trialkylboranes for the synthesis of adenosine A2A receptor antagonists, multicomponent C-alkylation reactions with aromatic aldimines, one-pot alkylation-boration and electrophilic substitution.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-56-5
Molecular Formula C12H27B
Molecular Weight (g/mol) 182.16
MDL Number MFCD00009423
InChI Key CMHHITPYCHHOGT-UHFFFAOYSA-N
Synonym tri-n-butylborane, borane, tributyl, tributylborine, tri-n-butyl borane, tributylboron, tri-n-butylboron, ccris 4133, solution, tributyl-borane, tributyl borane
PubChem CID 31216
IUPAC Name tributylborane
SMILES CCCCB(CCCC)CCCC

Specifications

Density 0.834
Quantity 500 mL
UN Number UN2924
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 182.16
Concentration or Composition (by Analyte or Components) 1M soln. in THF
Chemical Name or Material Tri-n-butylborane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.