accessibility menu, dialog, popup

UserName

CH3BCH3CH3

CAS RN 122-56-5

IUPAC Name: tributylborane
Synonyms: tri-n-butylborane
Molecular Weight (g/mol): 182.16
Molecular Formula: C12H27B
InChi Key: CMHHITPYCHHOGT-UHFFFAOYSA-N
SMILES: CCCCB(CCCC)CCCC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results