missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-n-butylborane, 1M soln. in THF, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tri-n-butylborane is used as a reactant for addition reactions for synthesis of phenyltrialkylborate salts, boron-based multicomponent reactions for generation of substituted aziridines, oxazolidines, and pyrrolidines, Suzuki-Miyaura cross coupling of halopurines with trialkylboranes for the synthesis of adenosine A2A receptor antagonists, multicomponent C-alkylation reactions with aromatic aldimines, one-pot alkylation-boration and electrophilic substitution.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Tri-n-butylborane is used as a reactant for addition reactions for synthesis of phenyltrialkylborate salts, boron-based multicomponent reactions for generation of substituted aziridines, oxazolidines, and pyrrolidines, Suzuki-Miyaura cross coupling of halopurines with trialkylboranes for the synthesis of adenosine A2A receptor antagonists, multicomponent C-alkylation reactions with aromatic aldimines, one-pot alkylation-boration and electrophilic substitution.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 0.834 |
| Quantity | 100 mL |
| UN Number | UN2924 |
| Solubility Information | Not miscible or difficult to mix with water. |
| Formula Weight | 182.16 |
| Concentration or Composition (by Analyte or Components) | 1M soln. in THF |
| Chemical Name or Material | Tri-n-butylborane |
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?