missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREM2 Antibody (2B5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
£163.00 - £444.00
Specifications
| Antigen | TREM2 |
|---|---|
| Clone | 2B5 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1000, Flow Cytometry 2-5ug, ELISA 1:500-1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18267910
|
Novus Biologicals
NBP1-07101SS |
0.02 mL |
£163.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18706303
|
Novus Biologicals
NBP1-07101 |
0.1 mL |
£444.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TREM2 Monoclonal specifically detects TREM2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TREM2 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Alzheimers Research, Immunology, Inflammation, Innate Immunity, Neuroscience | |
| Q9NZC2 | |
| 54209 | |
| Recombinant human TREM2 (19-161aa) purified from E. coli (NP_061838). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 2B5 | |
| Western Blot 1:500-1000, Flow Cytometry 2-5ug, ELISA 1:500-1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
| TREM2 | |
| IgG3 κ | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title